Assay Details
| Gene Symbol: Exosc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138586.3 | NP_613052.1 | 2 - 3 | 348 | 57 | |
| GenBank mRNA | AK006475.1 | - | 2 - 3 | 301 | 57 | |
| AK007372.1 | - | 2 - 3 | 337 | 57 | ||
| AK137780.1 | - | 311 | 57 | |||
| AK155228.1 | - | 2 - 3 | 300 | 57 | ||
| BB856072.1 | - | 2 - 3 | 350 | 57 | ||
| BC034358.1 | - | 2 - 3 | 294 | 2651418 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
rRNA processing
RNA catabolic process
rRNA catabolic process
rRNA 3'-end processing
nuclear-transcribed mRNA catabolic process, exonucleolytic, 3'-5'
U4 snRNA 3'-end processing
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
DNA deamination
defense response to virus
nuclear mRNA surveillance
polyadenylation-dependent snoRNA 3'-end processing
molecular_function
RNA binding
rRNA processing
RNA catabolic process
rRNA catabolic process
rRNA 3'-end processing
nuclear-transcribed mRNA catabolic process, exonucleolytic, 3'-5'
U4 snRNA 3'-end processing
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
DNA deamination
defense response to virus
nuclear mRNA surveillance
polyadenylation-dependent snoRNA 3'-end processing
molecular_function
RNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25011170 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25011168 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25011169 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440443 | BC092393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440443 | BE854983.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440443 | AK006475.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440443 | AK007372.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440443 | NM_138586.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440443 | BC092393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440443 | BE854983.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440443 | AK006475.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440443 | AK007372.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440443 | NM_138586.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669688 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 455313 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 162261 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 297966 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473923 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 865021 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 66934 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142925 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440443 | BC092393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440443 | BE854983.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440443 | AK006475.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440443 | AK007372.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440443 | NM_138586.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440443 | BC092393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440443 | BE854983.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440443 | AK006475.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440443 | AK007372.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440443 | NM_138586.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440443 | BC092393.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440443 | BE854983.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440443 | AK006475.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440443 | AK007372.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440443 | NM_138586.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440443 | BC092393.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440443 | BE854983.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440443 | AK006475.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440443 | AK007372.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440443 | NM_138586.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011180 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142925 | - | - |
| Whole Mouse Genome G4122A | A_51_P440443 | BC092393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440443 | BE854983.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440443 | AK006475.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440443 | AK007372.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440443 | NM_138586.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440443 | BC092393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440443 | BE854983.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440443 | AK006475.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440443 | AK007372.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440443 | NM_138586.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P963329 | AK086094.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P963329 | AK143763.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P963329 | AK053661.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134655 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142925 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06466333 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06466330 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25011170 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25011168 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25011169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06359792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06227678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06443352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05926826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06380424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06466330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06377641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06567267 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10551177 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551179 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551180 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551178 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551174 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551176 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551175 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417167_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10551173 | - | - |
| Mouse Genome 430 2.0 Array | 1417167_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417167_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27998_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27998_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700003292.mm.1 | TC0700000421.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700003292.mm.1 | NM_138586.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700003292.mm.1 | NM_138586.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700003292.mm.1 | NR_104358.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700003294.mm.1 | TC0700000421.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700003294.mm.1 | NM_138586.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700003294.mm.1 | NM_138586.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700003294.mm.1 | NR_104358.1 | Refseq |
| Mouse Expression Set 430 | 1417167_at | - | - |
| Mouse Expression Set 430 | 1417167_PM_at | - | - |
| Murine Genome U74v2 Set | 162153_i_at | - | - |
| Murine Genome U74v2 Set | 162153_PM_i_at | - | - |
| Murine Genome U74v2 Set | 97249_PM_at | - | - |
| Murine Genome U74v2 Set | 97249_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417167_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417167_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417167_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417167_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 907415 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3780064 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003780064 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2663498 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2663498 | - | - |
| mouse-6 v1.1 (Illumina) | scl32923.6.1_33-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3780064 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32923.6.1_33-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3780064 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2877280 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2877280 | - | - |

Exosc5
Genomic Map