Assay Details
| Gene Symbol: Gfm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138591.2 | NP_613057.2 | 16 - 17 | 2189 | 78 | |
| GenBank mRNA | AF315511.1 | - | 16 - 17 | 2156 | 78 | |
| BC013093.1 | - | 16 - 17 | 2192 | 3584730 | 78 | |
| BC031772.1 | - | 16 - 17 | 2156 | 4239980 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gfm1
Genomic Map