Assay Details
| Gene Symbol: Tomm70a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138599.5 | NP_613065.2 | 3 - 4 | 786 | 91 | |
| GenBank mRNA | AK012084.1 | - | 3 - 4 | 787 | 91 | |
| AK083586.1 | - | 3 - 4 | 787 | 91 | ||
| AK122356.1 | - | 719 | 91 | |||
| AK145458.1 | - | 3 - 4 | 799 | 91 | ||
| AK150429.1 | - | 3 - 4 | 787 | 91 | ||
| AK161720.1 | - | 3 - 4 | 776 | 91 | ||
| AK169809.1 | - | 3 - 4 | 749 | 91 | ||
| BC057096.1 | - | 3 - 4 | 787 | 5709110 | 91 | |
| BC139420.1 | - | 3 - 4 | 759 | 8862442 | 91 | |
| BC139421.1 | - | 3 - 4 | 759 | 8862443 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tomm70a
Genomic Map