Assay Details
| Gene Symbol: Tomm70a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138599.5 | NP_613065.2 | 7 - 8 | 1379 | 84 | |
| GenBank mRNA | AK012084.1 | - | 7 - 8 | 1380 | 84 | |
| AK083586.1 | - | 7 - 8 | 1380 | 84 | ||
| AK122356.1 | - | 1312 | 84 | |||
| AK145458.1 | - | 7 - 8 | 1392 | 84 | ||
| AK150429.1 | - | 7 - 8 | 1380 | 84 | ||
| AK161720.1 | - | 7 - 8 | 1369 | 84 | ||
| AK169809.1 | - | 7 - 8 | 1341 | 84 | ||
| BC057096.1 | - | 7 - 8 | 1380 | 5709110 | 84 | |
| BC139420.1 | - | 7 - 8 | 1352 | 8862442 | 84 | |
| BC139421.1 | - | 7 - 8 | 1352 | 8862443 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tomm70a
Genomic Map