Assay Details
| Gene Symbol: Ppp1r3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290574.1 | NP_001277503.1 | 2 - 3 | 1089 | 63 | |
| NM_138605.3 | NP_613071.3 | 2 - 3 | 1089 | 63 | ||
| GenBank mRNA | AF229644.1 | - | 2 - 3 | 928 | 63 | |
| BC059275.1 | - | 2 - 3 | 1023 | 63 | ||
| BC138137.1 | - | 2 - 3 | 921 | 8861156 | 63 | |
| BC138138.1 | - | 2 - 3 | 921 | 8861157 | 63 | |
More Information
Set Membership: |
|

Ppp1r3f
Genomic Map