| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00015201.mm.1 |
NM_001162423.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015201.mm.1 |
NM_001162423.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015201.mm.1 |
NM_001162424.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015201.mm.1 |
NM_001162424.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015201.mm.1 |
NM_138630.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015201.mm.1 |
NM_138630.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015201.mm.1 |
TC0X00003430.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00015200.mm.1 |
NM_001162423.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015200.mm.1 |
NM_001162423.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015200.mm.1 |
NM_001162424.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015200.mm.1 |
NM_001162424.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015200.mm.1 |
NM_138630.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015200.mm.1 |
NM_138630.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015200.mm.1 |
TC0X00003430.mm.1 |
MTA TC |