Assay Details
| Gene Symbol: Arhgap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162423.1 | NP_001155895.1 | 20 - 21 | 2582 | 82 | |
| NM_001162424.1 | NP_001155896.1 | 19 - 20 | 2552 | 82 | ||
| NM_138630.2 | NP_619536.2 | 20 - 21 | 2615 | 82 | ||
| GenBank mRNA | AF443826.1 | - | 20 - 21 | 2546 | 82 | |
| AK040687.1 | - | 13 - 14 | 1421 | 82 | ||
| AK129060.1 | - | 3124 | 82 | |||
| AK157294.1 | - | 20 - 21 | 2614 | 82 | ||
| AK169865.1 | - | 17 - 18 | 2050 | 82 | ||
| AY183124.1 | - | 20 - 21 | 2588 | 82 | ||
| AY183125.1 | - | 19 - 20 | 2525 | 82 | ||
| AY183126.1 | - | 19 - 20 | 2682 | 82 | ||
| AY183127.1 | - | 20 - 21 | 2588 | 82 | ||
| AY183128.1 | - | 16 - 17 | 2277 | 82 | ||
| AY183130.1 | - | 16 - 17 | 1960 | 82 | ||
| AY183131.1 | - | 15 - 16 | 2054 | 82 | ||
| AY183132.1 | - | 14 - 15 | 2280 | 82 | ||
| AY183133.1 | - | 18 - 19 | 2621 | 82 | ||
| BC109001.1 | - | 20 - 21 | 2513 | 40048081 | 82 | |
| BC109002.1 | - | 18 - 19 | 2572 | 82 | ||
More Information
Set Membership: |
|

Arhgap4
Genomic Map