Assay Details
| Gene Symbol: Mvd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138656.2 | NP_619597.2 | 2 - 3 | 200 | 109 | |
| GenBank mRNA | AJ309922.1 | - | 2 - 3 | 157 | 109 | |
| AK089354.1 | - | 2 - 3 | 202 | 109 | ||
| AK134803.1 | - | 2 - 3 | 189 | 109 | ||
| AK154883.1 | - | 2 - 3 | 202 | 109 | ||
| AK170007.1 | - | 2 - 3 | 137 | 109 | ||
| AK170773.1 | - | 2 - 3 | 196 | 109 | ||
| BC008526.1 | - | 2 - 3 | 168 | 3495424 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mvd
Genomic Map