Assay Details
| Gene Symbol: Socs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138657.3 | NP_619598.1 | 4 - 5 | 1151 | 99 | |
| GenBank mRNA | AF424814.1 | - | 4 - 5 | 1152 | 99 | |
| AK147580.1 | - | 4 - 5 | 1249 | 99 | ||
| AK147650.1 | - | 4 - 5 | 1249 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Socs7
Genomic Map