Assay Details
| Gene Symbol: Ufsp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138668.2 | NP_619609.1 | 11 - 12 | 1385 | 119 | |
| GenBank mRNA | AK075795.1 | - | 11 - 12 | 1380 | 119 | |
| AK078309.1 | - | 11 - 12 | 1386 | 119 | ||
| AK160164.1 | - | 10 - 11 | 1460 | 119 | ||
| BC005503.1 | - | 11 - 12 | 1386 | 3490744 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25833587 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245454 | AK160164.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245454 | AK075795.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245454 | BC005503.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245454 | NM_138668.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245454 | AK160164.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245454 | AK075795.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245454 | BC005503.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245454 | NM_138668.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 961305 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579042 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 296183 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947097 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 867075 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 341331 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925511 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454206 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 742196 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211391 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 421141 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 95964 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 711014 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 244176 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 934614 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35569 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119178 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120086 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166355 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245454 | AK160164.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245454 | AK075795.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245454 | BC005503.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245454 | NM_138668.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245454 | AK160164.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245454 | AK075795.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245454 | BC005503.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245454 | NM_138668.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25833590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25833614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25833606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25833605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25833607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25833603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25833588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25833617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25833589 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25833598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25833595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25833594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25833592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25833612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25833600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25833602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25833597 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25833610 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01727567 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01727575 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166355 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06498938 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P120086 | - | - |
| Whole Mouse Genome G4122A | A_51_P245454 | AK160164.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P245454 | AK075795.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P245454 | BC005503.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P245454 | NM_138668.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P245454 | AK160164.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P245454 | AK075795.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P245454 | BC005503.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P245454 | NM_138668.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P282302 | AK046862.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106003 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130061 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110248 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166355 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P120086 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06953995 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25833587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25833697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07233555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07234511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06883168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07073101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07195049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06693322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833595 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06683658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07007108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06954001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25833695 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10571622 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571627 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571625 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571624 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571630 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571629 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571628 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571633 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571626 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571623 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571632 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571631 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418319_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10571621 | - | - |
| Mouse Genome 430 2.0 Array | 1418319_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418319_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 192169_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 192169_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800003684.mm.1 | NM_138668.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800003684.mm.1 | TC0800000525.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800003687.mm.1 | NM_138668.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800003687.mm.1 | TC0800000525.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418319_PM_at | - | - |
| Mouse Expression Set 430 | 1418319_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418319_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418319_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418319_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418319_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 308387 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3390273 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1050154 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001050154 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003390273 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2717570 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257362 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257362 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2717570 | - | - |
| mouse-6 v1.1 (Illumina) | scl33851.12.1_1-S | - | - |
| mouse-6 v1.1 (Illumina) | scl000603.1_82-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1050154 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3390273 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33851.12.1_1-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000603.1_82-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1050154 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3390273 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2717569 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2957904 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2957898 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2717569 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2957898 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2957904 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl33851.12.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl000603.1_82-S | - | - |

Ufsp2
Genomic Map