Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200043558.mm.1 |
NM_138685.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043558.mm.1 |
NM_138685.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043558.mm.1 |
TC0200005108.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200043562.mm.1 |
NM_001045554.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043562.mm.1 |
NM_001045554.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043562.mm.1 |
NM_138685.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043562.mm.1 |
NM_138685.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043562.mm.1 |
TC0200005108.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200043557.mm.1 |
NM_001045554.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043557.mm.1 |
NM_001045554.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043557.mm.1 |
NM_138685.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043557.mm.1 |
NM_138685.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043557.mm.1 |
TC0200005108.mm.1 |
MTA TC |