Assay Details
Gene Symbol: Smim11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138743.2 | NP_620082.1 | 2 - 3 | 157 | 86 | |
GenBank mRNA | AK004538.1 | - | 2 - 3 | 157 | 86 | |
AY033901.1 | - | 2 - 3 | 152 | 86 | ||
BC005668.1 | - | 2 - 3 | 141 | 3152735 | 86 | |
BM932344.1 | - | 2 - 3 | 141 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK004538.1, AY033901.1, BC005668.1, NM_138743.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
