Assay Details
| Gene Symbol: Prom2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138750.2 | NP_620089.2 | 6 - 7 | 876 | 68 | |
| NM_178047.4 | NP_835148.2 | 6 - 7 | 876 | 68 | ||
| GenBank mRNA | AF269062.1 | - | 6 - 7 | 926 | 68 | |
| AK029851.1 | - | 6 - 7 | 857 | 68 | ||
| AK137749.1 | - | 6 - 7 | 854 | 68 | ||
| BC039212.1 | - | 6 - 7 | 844 | 4188940 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029851.1 transcripts.
Set Membership: |
|

Prom2
Genomic Map