Assay Details
| Gene Symbol: Ttc36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138951.1 | NP_620401.1 | 2 - 3 | 432 | 84 | |
| GenBank mRNA | AK149384.1 | - | 2 - 3 | 463 | 84 | |
| AK165811.1 | - | 2 - 3 | 330 | 84 | ||
| BC021608.1 | - | 2 - 3 | 344 | 5099426 | 84 | |
| BC021613.1 | - | 2 - 3 | 438 | 5102393 | 84 | |
| BC024432.1 | - | 2 - 3 | 432 | 4972715 | 84 | |
| BC026825.1 | - | 2 - 3 | 379 | 4240199 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26469623 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26469624 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26469625 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P198675 | BC021613.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P198675 | BC024432.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P198675 | BC026825.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P198675 | NM_138951.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P198675 | AK149384.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P198675 | BC021613.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P198675 | BC024432.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P198675 | BC026825.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P198675 | NM_138951.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P198675 | AK149384.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 286262 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 230510 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 59910 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136208 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P198675 | BC021613.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P198675 | BC024432.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P198675 | BC026825.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P198675 | NM_138951.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P198675 | AK149384.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P198675 | BC021613.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P198675 | BC024432.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P198675 | BC026825.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P198675 | NM_138951.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P198675 | AK149384.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469694 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26469677 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136208 | - | - |
| Whole Mouse Genome G4122A | A_51_P198675 | BC021613.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P198675 | BC024432.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P198675 | BC026825.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P198675 | NM_138951.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P198675 | AK149384.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P198675 | BC021613.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P198675 | BC024432.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P198675 | BC026825.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P198675 | NM_138951.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P198675 | AK149384.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120284 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136208 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07693046 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07693038 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07858826 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07693033 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07858843 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07858841 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07858835 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26469623 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26469624 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26469625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26469623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26469621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26469654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26469633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07693046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07858840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26469647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26469643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26469651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26469623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26469649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26469640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26469646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26469625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26469639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26469653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26469627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26469629 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10592941 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592940 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592939 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418696_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10592938 | - | - |
| Mouse Genome 430 2.0 Array | 1418696_at | - | - |
| Mouse Genome 430 2.0 Array | 1418696_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 192653_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 192653_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900018135.mm.1 | NM_138951.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900018135.mm.1 | NM_138951.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900018135.mm.1 | TC0900002180.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418696_PM_at | - | - |
| Mouse Expression Set 430 | 1418696_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418696_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418696_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418696_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418696_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 908221 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2190463 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002190463 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2670057 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2670057 | - | - |
| mouse-6 v1.1 (Illumina) | scl35941.3.1_31-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2190463 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl35941.3.1_31-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2190463 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2670057 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2670057 | - | - |

Ttc36
Genomic Map