Assay Details
| Gene Symbol: Ogt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290535.1 | NP_001277464.1 | 4 - 5 | 568 | 126 | |
| NM_139144.4 | NP_631883.2 | 4 - 5 | 765 | 126 | ||
| GenBank mRNA | AF363030.1 | - | 4 - 5 | 530 | 126 | |
| BC057319.1 | - | 4 - 5 | 688 | 6830845 | 126 | |
| CB520299.1 | - | 4 - 5 | 515 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ogt
Genomic Map