Assay Details
| Gene Symbol: Hlcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139145.4 | NP_631884.1 | 7 - 8 | 1842 | 68 | |
| GenBank mRNA | AB066227.1 | - | 6 - 7 | 1702 | 68 | |
| AK052424.1 | - | 8 - 9 | 2138 | 68 | ||
| AK158212.1 | - | 8 - 9 | 1959 | 68 | ||
| AK168925.1 | - | 4 - 5 | 737 | 68 | ||
| BC050090.1 | - | 7 - 8 | 1843 | 30012868 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hlcs
Genomic Map