Assay Details
| Gene Symbol: Cdkal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018346.1 | - | 7 - 8 | 637 | 78 | |
| AK041864.1 | - | 7 - 8 | 633 | 78 | ||
| AK082629.1 | - | 7 - 8 | 640 | 78 | ||
| AK158546.1 | - | 7 - 8 | 630 | 78 | ||
| AK169761.1 | - | 7 - 8 | 637 | 78 | ||
| BC016073.1 | - | 7 - 8 | 581 | 4482130 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdkal1
Genomic Map