Assay Details
| Gene Symbol: Lgi3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145219.4 | NP_660254.1 | 2 - 3 | 517 | 95 | |
| GenBank mRNA | AF515590.1 | - | 2 - 3 | 499 | 95 | |
| AK047495.1 | - | 2 - 3 | 469 | 95 | ||
| AK049831.1 | - | 2 - 3 | 469 | 95 | ||
| AK087125.1 | - | 3 - 4 | 383 | 95 | ||
| AK132296.1 | - | 2 - 3 | 511 | 95 | ||
| AK163664.1 | - | 2 - 3 | 469 | 95 | ||
| AY174077.1 | - | 2 - 3 | 499 | 95 | ||
| BC055315.1 | - | 2 - 3 | 377 | 5686778 | 95 | |
| BC061460.1 | - | 2 - 3 | 377 | 5686778 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lgi3
Genomic Map