Assay Details
| Gene Symbol: Appl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145220.2 | NP_660255.1 | 18 - 19 | 1748 | 86 | |
| GenBank mRNA | AK160075.1 | - | 18 - 19 | 1727 | 86 | |
| AK160105.1 | - | 18 - 19 | 1724 | 86 | ||
| AK168403.1 | - | 18 - 19 | 1711 | 86 | ||
| AK168748.1 | - | 18 - 19 | 1704 | 86 | ||
| AK171512.1 | - | 18 - 19 | 1704 | 86 | ||
| AY113706.1 | - | 18 - 19 | 1750 | 86 | ||
| BC002232.1 | - | 6 - 7 | 559 | 86 | ||
| BC048906.1 | - | 18 - 19 | 1686 | 6475576 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Appl2
Genomic Map