Assay Details
| Gene Symbol: Appl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145221.2 | NP_660256.1 | 17 - 18 | 1747 | 90 | |
| GenBank mRNA | AK030445.1 | - | 17 - 18 | 1626 | 90 | |
| AK033566.1 | - | 17 - 18 | 1718 | 90 | ||
| AK045438.1 | - | 17 - 18 | 1683 | 90 | ||
| AK049307.1 | - | 17 - 18 | 1734 | 90 | ||
| AK146356.1 | - | 17 - 18 | 1718 | 90 | ||
| AK173169.1 | - | 2 - 3 | 198 | 90 | ||
| AY113705.1 | - | 17 - 18 | 1712 | 90 | ||
| BC019708.1 | - | 9 - 10 | 1025 | 90 | ||
| BC063751.1 | - | 17 - 18 | 1759 | 6511401 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Appl1
Genomic Map