Assay Details
| Gene Symbol: Dhx35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291144.1 | NP_001278073.1 | 11 - 12 | 1107 | 68 | |
| NM_145742.2 | NP_665685.1 | 11 - 12 | 1087 | 68 | ||
| GenBank mRNA | AK004665.1 | - | 11 - 12 | 1033 | 68 | |
| AK155482.1 | - | 10 - 11 | 2511 | 68 | ||
| AK156955.1 | - | 11 - 12 | 1053 | 68 | ||
| AK170952.1 | - | 10 - 11 | 2511 | 68 | ||
| BC029709.1 | - | 11 - 12 | 1074 | 5359253 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhx35
Genomic Map