Assay Details
| Gene Symbol: Dhx35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291144.1 | NP_001278073.1 | 15 - 16 | 1603 | 78 | |
| NM_145742.2 | NP_665685.1 | 15 - 16 | 1583 | 78 | ||
| GenBank mRNA | AK004665.1 | - | 15 - 16 | 1529 | 78 | |
| AK155482.1 | - | 14 - 15 | 3009 | 78 | ||
| AK156955.1 | - | 15 - 16 | 1549 | 78 | ||
| AK170952.1 | - | 14 - 15 | 3007 | 78 | ||
| BC029709.1 | - | 15 - 16 | 1570 | 5359253 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhx35
Genomic Map