Assay Details
| Gene Symbol: Fads1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146094.2 | NP_666206.1 | 1 - 2 | 381 | 71 | |
| GenBank mRNA | AB072976.1 | - | 1 - 2 | 296 | 71 | |
| AF061504.1 | - | 1 - 2 | 283 | 71 | ||
| AK080706.1 | - | 1 - 2 | 285 | 71 | ||
| AK083959.1 | - | 1 - 2 | 243 | 71 | ||
| AK154367.1 | - | 1 - 2 | 331 | 71 | ||
| BC022139.1 | - | 1 - 2 | 326 | 5100199 | 71 | |
| BC026831.1 | - | 1 - 2 | 348 | 4239695 | 71 | |
| BC026848.1 | - | 1 - 2 | 321 | 4196030 | 71 | |
| BC063053.1 | - | 1 - 2 | 241 | 5696722 | 71 | |
| KF987079.1 | - | 1 - 2 | 265 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fads1
Genomic Map