Assay Details
| Gene Symbol: Fads1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146094.2 | NP_666206.1 | 2 - 3 | 483 | 61 | |
| GenBank mRNA | AB072976.1 | - | 2 - 3 | 398 | 61 | |
| AF061504.1 | - | 2 - 3 | 61 | |||
| AK080706.1 | - | 2 - 3 | 387 | 61 | ||
| AK083959.1 | - | 2 - 3 | 345 | 61 | ||
| AK154367.1 | - | 2 - 3 | 433 | 61 | ||
| BC022139.1 | - | 2 - 3 | 428 | 5100199 | 61 | |
| BC026831.1 | - | 2 - 3 | 450 | 4239695 | 61 | |
| BC026848.1 | - | 2 - 3 | 423 | 4196030 | 61 | |
| BC063053.1 | - | 2 - 3 | 343 | 5696722 | 61 | |
| KF987079.1 | - | 2 - 3 | 367 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fads1
Genomic Map