Assay Details
| Gene Symbol: Fads1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146094.2 | NP_666206.1 | 7 - 8 | 1057 | 93 | |
| GenBank mRNA | AB072976.1 | - | 7 - 8 | 972 | 93 | |
| AF061504.1 | - | 7 - 8 | 959 | 93 | ||
| AK033308.1 | - | 5 - 6 | 414 | 93 | ||
| AK083959.1 | - | 7 - 8 | 919 | 93 | ||
| AK154367.1 | - | 7 - 8 | 1007 | 93 | ||
| BC022139.1 | - | 7 - 8 | 5100199 | 93 | ||
| BC026831.1 | - | 7 - 8 | 4239695 | 93 | ||
| BC063053.1 | - | 7 - 8 | 917 | 5696722 | 93 | |
| KF987079.1 | - | 7 - 8 | 941 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fads1
Genomic Map