Assay Details
| Gene Symbol: Slc9a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB089793.1 | - | 14 - 15 | 1513 | 65 | |
| AF482993.1 | - | 13 - 14 | 1477 | 65 | ||
| AK049410.1 | - | 14 - 15 | 1509 | 65 | ||
| AK051791.1 | - | 13 - 14 | 1416 | 65 | ||
| AK129013.1 | - | 5 - 6 | 730 | 65 | ||
| AK143281.1 | - | 14 - 15 | 1482 | 65 | ||
| AK160826.1 | - | 11 - 12 | 1239 | 65 | ||
| AK173069.1 | - | 14 - 15 | 1849 | 65 | ||
| BC030879.1 | - | 14 - 15 | 1512 | 4241988 | 65 | |
| BC034508.1 | - | 7 - 8 | 913 | 65 | ||
| BC058947.1 | - | 14 - 15 | 1483 | 5702781 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a8
Genomic Map