Assay Details
| Gene Symbol: Slc44a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199186.1 | NP_001186115.1 | 11 - 12 | 1047 | 79 | |
| NM_152808.3 | NP_690021.1 | 11 - 12 | 1049 | 79 | ||
| GenBank mRNA | AK041533.1 | - | 11 - 12 | 1006 | 79 | |
| AK048648.1 | - | 11 - 12 | 1034 | 79 | ||
| AK153992.1 | - | 11 - 12 | 1028 | 79 | ||
| AK166695.1 | - | 11 - 12 | 1050 | 79 | ||
| BC013213.1 | - | 4 - 5 | 442 | 79 | ||
| BC031535.1 | - | 11 - 12 | 1025 | 3991609 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc44a2
Genomic Map