Assay Details
| Gene Symbol: Usp45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290425.1 | NP_001277354.1 | 3 - 4 | 331 | 64 | |
| NM_152825.2 | NP_690038.1 | 3 - 4 | 331 | 64 | ||
| GenBank mRNA | AK040660.1 | - | 3 - 4 | 332 | 64 | |
| AK088073.1 | - | 3 - 4 | 314 | 64 | ||
| AK162825.1 | - | 3 - 4 | 332 | 64 | ||
| BB213271.2 | - | 3 - 4 | 343 | 64 | ||
| BC027768.1 | - | 3 - 4 | 314 | 5354502 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp45
Genomic Map