Assay Details
| Gene Symbol: Usp45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290425.1 | NP_001277354.1 | 14 - 15 | 1984 | 93 | |
| NM_152825.2 | NP_690038.1 | 15 - 16 | 2128 | 93 | ||
| GenBank mRNA | AK019756.1 | - | 16 - 17 | 2243 | 93 | |
| AK036903.1 | - | 2 - 3 | 544 | 93 | ||
| AK088073.1 | - | 14 - 15 | 1967 | 93 | ||
| BC027768.1 | - | 15 - 16 | 2111 | 5354502 | 93 | |
| BM227454.2 | - | 3 - 4 | 93 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Usp45
Genomic Map