Assay Details
| Gene Symbol: Adcy8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291903.1 | NP_001278832.1 | 9 - 10 | 3395 | 99 | |
| NM_009623.2 | NP_033753.2 | 9 - 10 | 3395 | 99 | ||
| GenBank mRNA | AK138244.1 | - | 9 - 10 | 3396 | 99 | |
| BC059010.1 | - | 9 - 10 | 2492 | 99 | ||
| BC094231.1 | - | 9 - 10 | 2833 | 99 | ||
| U85021.1 | - | 9 - 10 | 3255 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adcy8
Genomic Map