Assay Details
| Gene Symbol: Angpt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009641.1 | NP_033771.1 | 3 - 4 | 618 | 102 | |
| GenBank mRNA | AF113707.1 | - | 3 - 4 | 618 | 102 | |
| AK132988.1 | - | 743 | 102 | |||
| AK154379.1 | - | 743 | 102 | |||
| BC129965.1 | - | 3 - 4 | 618 | 40106226 | 102 | |
| BC129966.1 | - | 3 - 4 | 618 | 40106227 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Angpt4
Genomic Map