Assay Details
| Gene Symbol: Alox15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009660.3 | NP_033790.3 | 9 - 10 | 1279 | 88 | |
| GenBank mRNA | AK142371.1 | - | 9 - 10 | 1280 | 88 | |
| BC056625.1 | - | 9 - 10 | 1266 | 5005087 | 88 | |
| BC081546.1 | - | 9 - 10 | 1265 | 3671948 | 88 | |
| CT010263.1 | - | 9 - 10 | 1253 | 88 | ||
| L34570.1 | - | 9 - 10 | 1263 | 88 | ||
| U04331.1 | - | 9 - 10 | 1253 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alox15
Genomic Map