Assay Details
| Gene Symbol: Agr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011783.2 | NP_035913.1 | 1 - 2 | 45 | 73 | |
| GenBank mRNA | AB016592.1 | - | 1 - 2 | 44 | 73 | |
| AF044262.1 | - | 1 - 2 | 45 | 73 | ||
| AK007677.1 | - | 1 - 2 | 64 | 73 | ||
| BC013334.1 | - | 1 - 2 | 63 | 4218549 | 73 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB016592.1,AF044262.1,AK007677.1,BC013334.1,NM_011783.2 transcripts.
Set Membership: |
|

Agr2
Genomic Map