Assay Details
| Gene Symbol: Anxa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136089.2 | NP_001129561.1 | 5 - 6 | 570 | 80 | |
| GenBank mRNA | AJ238978.1 | - | 5 - 6 | 470 | 80 | |
| AK008772.1 | - | 5 - 6 | 493 | 80 | ||
| AK008808.1 | - | 5 - 6 | 494 | 80 | ||
| AK018597.1 | - | 5 - 6 | 527 | 80 | ||
| AK131880.1 | - | 5 - 6 | 490 | 80 | ||
| AK168715.1 | - | 5 - 6 | 474 | 80 | ||
| BC125319.1 | - | 5 - 6 | 442 | 40129834 | 80 | |
| BC132207.1 | - | 5 - 6 | 442 | 40130484 | 80 | |
More Information
Set Membership: |
|

Anxa10
Genomic Map