Assay Details
| Gene Symbol: Anxa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136089.2 | NP_001129561.1 | 10 - 11 | 946 | 116 | |
| NM_011922.3 | NP_036052.2 | 9 - 10 | 886 | 116 | ||
| GenBank mRNA | AJ238978.1 | - | 10 - 11 | 846 | 116 | |
| AK008808.1 | - | 10 - 11 | 870 | 116 | ||
| AK020288.1 | - | 9 - 10 | 811 | 116 | ||
| AK131880.1 | - | 10 - 11 | 866 | 116 | ||
| AK168715.1 | - | 10 - 11 | 850 | 116 | ||
| BC125319.1 | - | 10 - 11 | 818 | 40129834 | 116 | |
| BC132207.1 | - | 10 - 11 | 818 | 40130484 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Anxa10
Genomic Map