Assay Details
| Gene Symbol: Angptl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011923.4 | NP_036053.2 | 2 - 3 | 1346 | 64 | |
| GenBank mRNA | AF125176.1 | - | 1 - 2 | 833 | 64 | |
| AK037265.1 | - | 2 - 3 | 1239 | 64 | ||
| AK155464.1 | - | 2 - 3 | 1347 | 64 | ||
| BC138609.1 | - | 2 - 3 | 1152 | 8861629 | 64 | |
| BC138610.1 | - | 2 - 3 | 1152 | 8861630 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Angptl2
Genomic Map