Assay Details
| Gene Symbol: Anxa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281845.1 | NP_001268774.1 | 4 - 5 | 396 | 92 | |
| NM_013473.4 | NP_038501.2 | 4 - 5 | 396 | 92 | ||
| GenBank mRNA | AJ002390.1 | - | 4 - 5 | 372 | 92 | |
| AK009085.1 | - | 4 - 5 | 358 | 92 | ||
| AK075870.1 | - | 4 - 5 | 330 | 92 | ||
| BC030407.1 | - | 4 - 5 | 409 | 5322310 | 92 | |
More Information
Set Membership: |
|

Anxa8
Genomic Map