Assay Details
| Gene Symbol: Adat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013925.4 | NP_038953.1 | 2 - 3 | 328 | 66 | |
| GenBank mRNA | AF192375.1 | - | 1 - 2 | 167 | 66 | |
| AK035614.1 | - | 2 - 3 | 316 | 66 | ||
| AK039699.1 | - | 2 - 3 | 298 | 66 | ||
| AK082399.1 | - | 2 - 3 | 329 | 66 | ||
| AK161044.1 | - | 2 - 3 | 298 | 66 | ||
| BC019976.1 | - | 2 - 3 | 274 | 4458235 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adat1
Genomic Map