| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800022868.mm.1 |
NM_013925.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800022868.mm.1 |
NM_013925.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800022868.mm.1 |
TC0800002960.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800022869.mm.1 |
NM_013925.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800022869.mm.1 |
NM_013925.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800022869.mm.1 |
TC0800002960.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800022867.mm.1 |
NM_013925.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800022867.mm.1 |
NM_013925.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800022867.mm.1 |
TC0800002960.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800022863.mm.1 |
NM_013925.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800022863.mm.1 |
NM_013925.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800022863.mm.1 |
TC0800002960.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800022866.mm.1 |
TC0800002960.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800022870.mm.1 |
NM_013925.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800022870.mm.1 |
NM_013925.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800022870.mm.1 |
TC0800002960.mm.1 |
MTA TC |