Assay Details
| Gene Symbol: Agxt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276710.1 | NP_001263639.1 | 4 - 5 | 646 | 75 | |
| NM_016702.3 | NP_057911.2 | 4 - 5 | 646 | 75 | ||
| GenBank mRNA | AF027730.2 | - | 4 - 5 | 641 | 75 | |
| AK149432.1 | - | 4 - 5 | 569 | 75 | ||
| BC025799.1 | - | 4 - 5 | 610 | 5099627 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Agxt
Genomic Map