Assay Details
| Gene Symbol: Adar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038587.4 | NP_001033676.2 | 2 - 3 | 1561 | 87 | |
| NM_001146296.1 | NP_001139768.1 | 2 - 3 | 1553 | 87 | ||
| NM_019655.3 | NP_062629.3 | 2 - 3 | 1553 | 87 | ||
| GenBank mRNA | AF052506.1 | - | 2 - 3 | 1466 | 87 | |
| AF291050.1 | - | 2 - 3 | 1460 | 87 | ||
| AF291876.1 | - | 2 - 3 | 1460 | 87 | ||
| AK076413.1 | - | 2 - 3 | 1512 | 87 | ||
| AK089451.1 | - | 2 - 3 | 1524 | 87 | ||
| AK147614.1 | - | 2 - 3 | 1524 | 87 | ||
| AK156093.1 | - | 1 - 2 | 120 | 87 | ||
| AK172523.1 | - | 1 - 2 | 120 | 87 | ||
| BC042505.1 | - | 2 - 3 | 1487 | 4012994 | 87 | |
| CT010323.1 | - | 2 - 3 | 1460 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adar
Genomic Map