Assay Details
| Gene Symbol: Anapc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042491.2 | NP_001035956.1 | 7 - 8 | 1099 | 113 | |
| NM_001289517.1 | NP_001276446.1 | 7 - 8 | 1099 | 113 | ||
| NM_001289518.1 | NP_001276447.1 | 7 - 8 | 1099 | 113 | ||
| NM_001289519.1 | NP_001276448.1 | 7 - 8 | 1099 | 113 | ||
| NM_001289520.1 | NP_001276449.1 | 7 - 8 | 1099 | 113 | ||
| NM_021505.3 | NP_067480.2 | 7 - 8 | 1099 | 113 | ||
| GenBank mRNA | AB041593.1 | - | 7 - 8 | 1043 | 114 | |
| AK031432.1 | - | 7 - 8 | 1024 | 113 | ||
| AK049719.1 | - | 7 - 8 | 1053 | 113 | ||
| AK088327.1 | - | 7 - 8 | 1062 | 113 | ||
| AK147951.1 | - | 7 - 8 | 1038 | 113 | ||
| AK148679.1 | - | 7 - 8 | 1054 | 113 | ||
| AK151314.1 | - | 7 - 8 | 1045 | 113 | ||
| AK151905.1 | - | 7 - 8 | 1045 | 113 | ||
| AK152528.1 | - | 7 - 8 | 1015 | 113 | ||
| AK152981.1 | - | 7 - 8 | 1046 | 114 | ||
| AK153373.1 | - | 7 - 8 | 1045 | 113 | ||
| AK153422.1 | - | 7 - 8 | 1045 | 113 | ||
| AK159204.1 | - | 7 - 8 | 1045 | 113 | ||
| AK159713.1 | - | 7 - 8 | 1038 | 113 | ||
| AK159916.1 | - | 7 - 8 | 1038 | 113 | ||
| AK167561.1 | - | 7 - 8 | 1038 | 113 | ||
| AK168135.1 | - | 7 - 8 | 1023 | 113 | ||
| AK168737.1 | - | 7 - 8 | 1039 | 113 | ||
| AK168814.1 | - | 7 - 8 | 1038 | 113 | ||
| AK169196.1 | - | 7 - 8 | 1062 | 113 | ||
| BC010339.1 | - | 7 - 8 | 1015 | 2648173 | 113 | |
| BC046566.1 | - | 7 - 8 | 1078 | 2650790 | 113 | |
| BC046804.1 | - | 7 - 8 | 1036 | 6404923 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Anapc5
Genomic Map