Assay Details
| Gene Symbol: Krt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023256.1 | NP_075745.1 | 3 - 4 | 702 | 57 | |
| GenBank mRNA | AF473907.1 | - | 3 - 4 | 679 | 57 | |
| AK018567.1 | - | 3 - 4 | 702 | 57 | ||
| BC057172.1 | - | 3 - 4 | 685 | 4989154 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Krt20
Genomic Map