Assay Details
| Gene Symbol: Bmyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023326.2 | NP_075815.2 | 1 | 292 | 59 | |
| GenBank mRNA | AK012810.1 | - | 1 | 334 | 59 | |
| AK013473.1 | - | 1 | 342 | 59 | ||
| AK013780.1 | - | 1 | 339 | 59 | ||
| AK158603.1 | - | 1 | 342 | 59 | ||
| BC056436.1 | - | 1 | 292 | 6824373 | 59 | |
| BC060207.1 | - | 1 | 292 | 6824373 | 59 | |
| BC061001.1 | - | 1 | 391 | 6595436 | 59 | |
| Gene Symbol: A230005M16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK038413.1 | - | 1 | 59 | ||
| AK046328.1 | - | 1 | 59 | |||
| AK082064.1 | - | 2 | 59 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21073893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21073892 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21073895 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21073894 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P466853 | AK013473.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P466853 | AK013780.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P466853 | AK158603.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P466853 | BC056436.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P466853 | BC060207.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P466853 | AK013473.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P466853 | AK013780.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P466853 | AK158603.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P466853 | BC056436.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P466853 | BC060207.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165016 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P466853 | AK013473.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P466853 | AK013780.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P466853 | AK158603.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P466853 | BC056436.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P466853 | BC060207.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P466853 | AK013473.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P466853 | AK013780.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P466853 | AK158603.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P466853 | BC056436.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P466853 | BC060207.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073888 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165016 | - | - |
| Whole Mouse Genome G4122A | A_52_P466853 | AK013473.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P466853 | AK013780.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P466853 | AK158603.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P466853 | BC056436.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P466853 | BC060207.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P466853 | AK013473.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P466853 | AK013780.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P466853 | AK158603.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P466853 | BC056436.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P466853 | BC060207.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135786 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165016 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01201745 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01218423 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01218414 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01218424 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21073894 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21073892 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21073893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21073895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02165647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01638331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01949832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01638328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01638327 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01949794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01638326 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01949788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073938 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073891 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10470184 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470183 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428669_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10470182 | - | - |
| Mouse Genome 430 2.0 Array | 1428669_at | - | - |
| Mouse Genome 430 2.0 Array | 1428669_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107771_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107771_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200002661.mm.1 | NM_023326.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002661.mm.1 | NM_023326.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002661.mm.1 | TC0200000432.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200002660.mm.1 | TC0200000432.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428669_at | - | - |
| Mouse Expression Set 430 | 1428669_PM_at | - | - |
| Murine Genome U74v2 Set | 99514_at | - | - |
| Murine Genome U74v2 Set | 99514_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428669_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428669_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428669_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428669_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 405866 | - | - |
| ABI Mouse Genome Survey Microarray | 756275 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5270035 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005270035 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2776140 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2776140 | - | - |
| mouse-6 v1.1 (Illumina) | scl21165.1.12_85-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5270035 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21165.1.12_85-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270035 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2776140 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2776140 | - | - |

Bmyc
Genomic Map