Assay Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023617.2 | NP_076106.2 | 22 - 23 | 2587 | 67 | |
| GenBank mRNA | AF172276.2 | - | 22 - 23 | 2663 | 67 | |
| AK004700.1 | - | 11 - 12 | 1383 | 67 | ||
| AK030109.1 | - | 18 - 19 | 2127 | 67 | ||
| AK149434.1 | - | 22 - 23 | 2548 | 67 | ||
| AY347569.1 | - | 22 - 23 | 2464 | 67 | ||
| AY347570.1 | - | 22 - 23 | 2464 | 67 | ||
| BC049853.1 | - | 22 - 23 | 2539 | 67 | ||
| BC138876.1 | - | 22 - 23 | 2566 | 8861898 | 67 | |
More Information
Set Membership: |
|

Aox3
Genomic Map