Assay Details
| Gene Symbol: Rab22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024436.3 | NP_077756.2 | 1 - 2 | 300 | 66 | |
| GenBank mRNA | AB232616.1 | - | 1 - 2 | 39 | 66 | |
| AJ311124.1 | - | 1 - 2 | 96 | 66 | ||
| AK014407.1 | - | 286 | 66 | |||
| AK031667.1 | - | 1 - 2 | 280 | 66 | ||
| AK146594.1 | - | 1 - 2 | 300 | 66 | ||
| AK147619.1 | - | 386 | 66 | |||
| AK149200.1 | - | 292 | 66 | |||
| AK150617.1 | - | 1 - 2 | 306 | 66 | ||
| BC006596.1 | - | 1 - 2 | 287 | 3965264 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab22a
Genomic Map