Assay Details
| Gene Symbol: Derl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024440.2 | NP_077760.1 | 2 - 3 | 184 | 62 | |
| GenBank mRNA | AB047555.1 | - | 2 - 3 | 184 | 62 | |
| AK007348.1 | - | 2 - 3 | 169 | 62 | ||
| AK007948.1 | - | 2 - 3 | 170 | 62 | ||
| AK172429.1 | - | 2 - 3 | 170 | 62 | ||
| BC004729.1 | - | 2 - 3 | 167 | 62 | ||
| BC019815.1 | - | 2 - 3 | 163 | 62 | ||
| BC099615.1 | - | 2 - 3 | 164 | 62 | ||
| BC115466.1 | - | 2 - 3 | 179 | 40100387 | 62 | |
| BC115467.1 | - | 2 - 3 | 179 | 40100388 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Derl3
Genomic Map