Assay Details
| Gene Symbol: Derl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024440.2 | NP_077760.1 | 6 - 7 | 640 | 68 | |
| GenBank mRNA | AB047555.1 | - | 6 - 7 | 640 | 68 | |
| AK007348.1 | - | 6 - 7 | 502 | 68 | ||
| AK007948.1 | - | 6 - 7 | 626 | 68 | ||
| AK172429.1 | - | 6 - 7 | 626 | 68 | ||
| BC004729.1 | - | 6 - 7 | 623 | 68 | ||
| BC019815.1 | - | 6 - 7 | 620 | 68 | ||
| BC099615.1 | - | 6 - 7 | 620 | 68 | ||
| BC115466.1 | - | 6 - 7 | 635 | 40100387 | 68 | |
| BC115467.1 | - | 6 - 7 | 635 | 40100388 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27259217 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27259219 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27259221 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27259220 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27259218 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27259222 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P491667 | AB047555.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P491667 | AB047555.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 106310 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162832 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P491667 | AB047555.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P491667 | AB047555.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27259208 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162832 | - | - |
| Whole Mouse Genome G4122A | A_51_P491667 | AB047555.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P491667 | AB047555.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130372 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130609 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162832 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08250174 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08250135 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08250179 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08349718 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27259220 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27259221 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27259217 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27259218 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27259222 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27259219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27259233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08250148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08887645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08250135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08887657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08825604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08887656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08250173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08887647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08349718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08887649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08250140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08974160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27259183 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10364095 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364099 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364096 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364098 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364094 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364100 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364101 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364097 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453677_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10364093 | - | - |
| Mouse Genome 430 2.0 Array | 1453677_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1453677_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70377_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70377_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000005193.mm.1 | NM_024440.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005193.mm.1 | NM_024440.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005193.mm.1 | TC1000000726.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000005192.mm.1 | NM_024440.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005192.mm.1 | NM_024440.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005192.mm.1 | TC1000000726.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000005191.mm.1 | NM_024440.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005191.mm.1 | NM_024440.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005191.mm.1 | TC1000000726.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453677_a_at | - | - |
| Mouse Expression Set 430 | 1453677_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453677_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453677_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453677_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453677_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1690408 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1690408 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001690408 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1234931 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234931 | - | - |
| mouse-6 v1.1 (Illumina) | scl38758.5.1_28-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1690408 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38758.5.1_28-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1690408 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1234931 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234931 | - | - |

Derl3
Genomic Map