Assay Details
| Gene Symbol: Fam3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025473.3 | NP_079749.1 | 5 - 6 | 790 | 67 | |
| GenBank mRNA | AK007715.1 | - | 5 - 6 | 791 | 67 | |
| AK012353.1 | - | 1 - 2 | 118 | 67 | ||
| BC011237.1 | - | 5 - 6 | 788 | 67 | ||
| BC019752.1 | - | 6 - 7 | 556 | 67 | ||
| BC054411.1 | - | 5 - 6 | 462 | 67 | ||
| BC069871.1 | - | 5 - 6 | 925 | 6467541 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam3a
Genomic Map