Assay Details
| Gene Symbol: Rtca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025517.3 | NP_079793.2 | 6 - 7 | 804 | 82 | |
| GenBank mRNA | AK002543.1 | - | 6 - 7 | 714 | 82 | |
| AK009245.1 | - | 6 - 7 | 740 | 82 | ||
| AK151108.1 | - | 6 - 7 | 692 | 82 | ||
| AK151400.1 | - | 6 - 7 | 692 | 82 | ||
| AK152129.1 | - | 6 - 7 | 805 | 82 | ||
| AK159961.1 | - | 6 - 7 | 694 | 82 | ||
| AK167830.1 | - | 6 - 7 | 681 | 82 | ||
More Information
Set Membership: |
|

Rtca
Genomic Map